CAS RN 1594094-64-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CW-069, MedChemExpress
MedChemExpress CW-069 is an allosteric inhibitor of microtubule motor protein HSET with an IC50 of 75 μM.