CAS RN 159182-43-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
2
L755507, MedChemExpress
MedChemExpress L755507 is a potent, selective agonist of β3-AR with an IC50 of 35 nM. L755507 enhances the homology-directed repair (HDR)-mediated genome editing in CRISPR/Cas9 nickase system.