CAS RN 1589527-65-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
KIRA6, MedChemExpress
MedChemExpress KIRA6 is an advanced small-molecule IRE1α RNase kinase inhibitor with an IC50 of 0.6 μM. KIRA6 can trigger an apoptotic response.