CAS RN 1588521-78-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BET-BAY 002, MedChemExpress
MedChemExpress BET-BAY 002 is a potent BET inhibitor; shows efficacy in a multiple myeloma model.