CAS RN 1575596-29-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Rilzabrutinib, MedChemExpress
MedChemExpress Rilzabrutinib (PRN1008) is a reversible covalent, selective and oral active inhibitor of Bruton’s Tyrosine Kinase (BTK), with an IC50 of 1.3 nM.