CAS RN 155521-45-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Alisol F, MedChemExpress
MedChemExpress Alisol F is a triterpene isolated from Alisma orientalis, has immunosuppressive and anti-virus functions. Alisol F exhibits inhibitory activity in vitro on hepatitis B virus (HBV) surface antigen (HBsAg) secretion of the HepG2.2.15 cell line with an IC50 of 0.6 μM.