CAS RN 1514888-56-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
S1p receptor agonist 1, MedChemExpress
MedChemExpress S1p receptor agonist 1 is a potent and orally active S1P receptor agonist, exhibits an activity of inducing S1P1 internalization (EC50=9.83 nM). S1p receptor agonist 1 has the potential for the study of arthritis and EAE (experimental autoimmune encephalitis). S1p receptor agonist 1 is extracted from patent WO2015039587A1, Compound 2.