CAS RN 149385-65-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress β-Amyloid (31-35)
MedChemExpress β-Amyloid (31-35) is the shortest sequence of native Amyloid-β peptide that retains neurotoxic activity.
2
MedChemExpress β-Amyloid (31-35)
MedChemExpress β-Amyloid (31-35) is the shortest sequence of native Amyloid-β peptide that retains neurotoxic activity.
3
MedChemExpress β-Amyloid (31-35)
MedChemExpress β-Amyloid (31-35) is the shortest sequence of native Amyloid-β peptide that retains neurotoxic activity.