CAS RN 146949-21-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pocapavir, MedChemExpress
MedChemExpress Pocapavir (SCH-48973) is an orally active capsid inhibitor. Pocapavir prevents virion uncoating upon entry into the cell. Pocapavir has antiviral activity against polioviruses. Pocapavir also inhibits enterovirus infections.