CAS RN 1464151-33-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ETC-206, MedChemExpress
MedChemExpress ETC-206 is a selective MNK1 and MNK2 inhibitor with IC50s of 64 nM and 86 nM, respectively.