CAS RN 1454846-35-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Lorlatinib, MedChemExpress
MedChemExpress Lorlatinib (PF-06463922) is a selective, orally active, brain-penetrant and ATP-competitive ROS1/ALK inhibitor. Lorlatinib has Kis of L1196M, respectively. Lorlatinib has anticancer activity.