CAS RN 1453097-13-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HUHS015, MedChemExpress
MedChemExpress HUHS015 is a potent prostate cancer antigen-1 (PCA-1/ALKBH3) inhibitor with an IC50 of 0.67μM for PCA-1.