CAS RN 1450929-77-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LHF-535, MedChemExpress
MedChemExpress LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively.