CAS RN 1448671-31-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AZD3965, MedChemExpress
MedChemExpress AZD3965 is a selective MCT1 inhibitor with a Ki of 1.6 nM, showing 6-fold selectivity over MCT2.