CAS RN 1445993-26-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Mivebresib, MedChemExpress
MedChemExpress Mivebresib (ABBV-075) is a potent and orally active bromodomain and extraterminal domain (BET) bromodomain inhibitor. Mivebresib binds to BRD4 with a Ki of 1.5 nM.