CAS RN 1445179-97-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress D-JNKI-1
MedChemExpress D-JNKI-1 (AM-111) is a highly potent and cell-permeable peptide inhibitor of JNK.
2
MedChemExpress D-JNKI-1
MedChemExpress D-JNKI-1 (AM-111) is a highly potent and cell-permeable peptide inhibitor of JNK.
3
MedChemExpress D-JNKI-1
MedChemExpress D-JNKI-1 (AM-111) is a highly potent and cell-permeable peptide inhibitor of JNK.
4
MedChemExpress D-JNKI-1
MedChemExpress D-JNKI-1 (AM-111) is a highly potent and cell-permeable peptide inhibitor of JNK.