CAS RN 1443460-91-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK2838232, MedChemExpress
MedChemExpress GSK2838232 inhibit HIV reverse transcriptase activity across a broad panel of HIV-1 isolates, extracted from patent WO/2013090664A1, compound51.