CAS RN 1443138-53-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK2200150A, MedChemExpress
MedChemExpress GSK2200150A, identified by high-throughput screening (HTS) campaign, is an anti-tuberculosis (TB) agent.