CAS RN 1440898-61-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Deltarasin, MedChemExpress
MedChemExpress Deltarasin is an inhibitor of KRAS-PDEδ interaction with Kd of 38 nM for binding to purified PDEδ.