CAS RN 143901-35-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Aglafoline, MedChemExpress
MedChemExpress Aglafoline inhibits in a selective and concentration-dependent manner the aggregation and ATP release reaction induced in washed rabbit platelets by PAF (platelet-activating factor).