CAS RN 1436383-95-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ BF 738735
Potent and selective PI 4-kinase IIIβ inhibitor; broad spectrum enterovirus replication inhibitor
2
BF738735, MedChemExpress
MedChemExpress BF738735 is a phosphatidylinositol 4-kinase III beta (PI4KIIIβ) inhibitor with an IC50 of 5.7 nM.