CAS RN 143621-35-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
5
de
5
resultados
1
Tocris Bioscience™ ML 233
Non-peptide apelin receptor agonist
2
3-AP, MedChemExpress
MedChemExpress 3-AP (PAN-811) is a potent inhibitor of the M2 subunit of ribonucleotide reductase (RR), and is a potent radiosensitizer.
3
Tocris Bioscience™ 3-AP
Ribonucleotide reductase inhibitor; also an iron chelator
4
5