CAS RN 1435786-04-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Rapastinel Trifluoroacetate, MedChemExpress
MedChemExpress Rapastinel Trifluoroacetate (GLYX-13 Trifluoroacetate) is an NMDA receptor modulator with glycine-site partial agonist properties. Rapastinel Trifluoroacetate has the potential for major depressive disorder treatment.