CAS RN 14357-08-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
N6-Ethyladenosine, MedChemExpress
MedChemExpress N6-Ethyladenosine is an adenosine derivative, acts as a Adenosine receptor agonist, with Kis of 4.9 and 4.7 nM for hA1AR and hA3AR, respectively.