Learn More
N6-Ethyladenosine, MedChemExpress
Código de producto. 16460837
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16460837 50 mg 50 miligramos
16424538 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16460837 Proveedor Medchem Express N.º de proveedor HY111809
MedChemExpress N6-Ethyladenosine is an adenosine derivative, acts as a Adenosine receptor agonist, with Kis of 4.9 and 4.7 nM for hA1AR and hA3AR, respectively.
N6-Ethyladenosine is an adenosine derivative, acts as a Adenosine receptor agonist, with Kis of 4.9 and 4.7 nM for hA1AR and hA3AR, respectively.
Especificaciones
Nombre del producto químico o material N6-Ethyladenosine
CAS 14357-08-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C12H17N5O4
Cantidad 50 mg
Información de solubilidad DMSO : ≥ 83.33 mg/mL (282.20 mM)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCC)O1)O)O
Peso molecular (g/mol) 295.29
Formula Weight (peso de la fórmula) 295.29
Porcentaje de pureza 99.53%
Grado Research
Forma física Solid