CAS RN 1435488-37-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
XMD17-109, MedChemExpress
MedChemExpress XMD17-109 is a novel, specific ERK-5 inhibitor, with an IC50 of 162 nM.