CAS RN 1431612-23-5
IUPAC Nombre:
N-[(6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl]-1-(propan-2-yl)-6-{6-[4-(propan-2-yl)piperazin-1-yl]pyridin-3-yl}-1H-indazole-4-carboxamide
Sinónimos:
1-isopropyl-6-[6-(4-isopropylpiperazin-1-yl)pyridin-3-yl]-N-[(6-methyl-2-oxo-4-propyl-1H-pyridin-3-yl)methyl]indazole-4-carboxamide
Peso molecular (g/mol):
569.75
Fórmula molecular:
C33H43N7O2
InChi Key:
DPJNKUOXBZSZAI-UHFFFAOYSA-N
SMILES:
CCCC1=C(CNC(=O)C2=C3C=NN(C(C)C)C3=CC(=C2)C2=CN=C(C=C2)N2CCN(CC2)C(C)C)C(=O)NC(C)=C1
1
–
3
de
3
resultados
3
UNC1999, MedChemExpress
MedChemExpress UNC1999 is a SAM-competitive, potent and selective inhibitor of EZH2/1 with IC50s of <10 nM and 45 nM, repectively.