CAS RN 1430218-51-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
VI 16832, MedChemExpress
MedChemExpress VI 16832 is a broad spectrum Type I kinase inhibitor which can be used as an enrichment tool for the comparative expression analysis of protein kinases in different cancer cell lines.