CAS RN 1429882-07-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MRX-2843, TRC
CAS: 1429882-07-4 Fórmula molecular: C29H40N6O Peso molecular (g/mol): 488.67 Sinónimo: trans-4-[2-[(2-Cyclopropylethyl)amino]-5-[4-[(4-methyl-1-piperazinyl)methyl]phenyl]-7H-pyrrolo[2,3-d]pyrimidin-7-yl]cyclohexanol Nombre IUPAC: 4-[2-(2-ciclopropiletilamino)-5-[4-[(4-metilpiperazina-1-il)metil]fenil]pirrrolo[2,3-d]pirimidina-7-il]ciclohexan-1-ol SMILES: CN1CCN(CC2=CC=C(C=C2)C3=CN([C@H]4CC[C@H](O)CC4)C5=C3C=NC(NCCC6CC6)=N5)CC1
2
MRX-2843, MedChemExpress
MedChemExpress MRX-2843 (UNC2371) is an orally active, ATP-competitive dual MERTK and FLT3 tyrosine kinases inhibitor (TKI) with enzymatic IC50s of 1.3 nM for MERTK and 0.64 nM for FLT3, respectively.