CAS RN 1429054-28-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IDF-11774, MedChemExpress
MedChemExpress IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.