CAS RN 142344-87-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GP531, MedChemExpress
MedChemExpress GP531 is a potent, second-generation adenosine regulating agent, is pharmacologically silent under basal conditions but increases localized endogenous adenosine during ischemia.