CAS RN 1420071-30-2
IUPAC Nombre:
5-(5-{[3-(4-bromophenyl)-2-imino-4-oxo-1,3-thiazolidin-5-ylidene]methyl}furan-2-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Sinónimos:
5-(5-{[3-(4-bromophenyl)-2-imino-4-oxo-1,3-thiazolidin-5-ylidene]methyl}furan-2-yl)-2H-isoindole-1,3-dione
Peso molecular (g/mol):
494.32
Fórmula molecular:
C22H12BrN3O4S
InChi Key:
ULBOWKXOFOTCMU-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)N1C(=N)SC(=CC2=CC=C(O2)C2=CC3=C(C=C2)C(=O)NC3=O)C1=O
1
–
1
de
1
resultados
1
Bioymifi, MedChemExpress
MedChemExpress Bioymifi (DR5 Activator), a potent TRAIL receptor DR5 activator, binds to the extracellular domain (ECD) of DR5 with a Kd of 1.2 μM. Bioymifi can act as a single agent to induce DR5 clustering and aggregation, leading to apoptosis.