CAS RN 1416270-18-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Mutant IDH1-IN-4, MedChemExpress
MedChemExpress Mutant IDH1-IN-4 (compound 434) is an inhibitor of mutant Isocitrate dehydrogenase 1 (IDH 1), with IC50 values of ≤ 0.5 μM for mutant IDH1 in R132H, HT1080 and U87R132H cells.