CAS RN 1415560-64-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK256066 Trifluoroacetate, MedChemExpress
MedChemExpress GSK256066 Trifluoroacetate is a selective and high-affinity phosphodiesterase 4 (PDE) inhibitor, with an IC50 of 3.2 pM for PDE4B. GSK256066 Trifluoroacetate is developed for the research of chronic obstructive pulmonary disease.