Learn More
GSK256066 Trifluoroacetate, MedChemExpress
Código de producto. 16490628
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16490628 5 mg 5 miligramos
16484087 50 mg 50 miligramos
16478188 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16490628 Proveedor Medchem Express N.º de proveedor HY70069
MedChemExpress GSK256066 Trifluoroacetate is a selective and high-affinity phosphodiesterase 4 (PDE) inhibitor, with an IC50 of 3.2 pM for PDE4B. GSK256066 Trifluoroacetate is developed for the research of chronic obstructive pulmonary disease.
GSK256066 Trifluoroacetate is a selective and high-affinity phosphodiesterase 4 (PDE) inhibitor, with an IC50 of 3.2 pM for PDE4B. GSK256066 Trifluoroacetate is developed for the research of chronic obstructive pulmonary disease.
Especificaciones
Nombre del producto químico o material GSK256066 Trifluoroacetate
CAS 1415560-64-3
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C29H27F3N4O7S
Cantidad 5 mg
Información de solubilidad DMSO : 50 mg/mL (79.04 mM; Need ultrasonic)
SMILES OC(C(F)(F)F)=O.O=C(N)C1=C(C2=CC(S(=O)(C3=CC=CC(C(N(C)C)=O)=C3)=O)=CC(C)=C2N=C1)NC4=CC=CC(OC)=C4
Peso molecular (g/mol) 632.61
Formula Weight (peso de la fórmula) 632.61
Porcentaje de pureza 99.54%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation