CAS RN 1412458-61-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Asenapine hydrochloride, MedChemExpress
MedChemExpress Asenapine hydrochloride, an antipsychotic, is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and Dopamine (D2, D3, D4) receptor antagonist with Ki values of 0.03-4.0 nM for 5-HT and 1.3, 0.42, 1.1 nM for Dopamine receptor, respectively.