CAS RN 140896-21-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Indolicidin
MedChemExpress Indolicidin is a potent antimicrobial peptide purified from the cytoplasmic granules of bovine neutrophils.
2
MedChemExpress Indolicidin
MedChemExpress Indolicidin is a potent antimicrobial peptide purified from the cytoplasmic granules of bovine neutrophils.