CAS RN 1402837-79-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IDO-IN-5, MedChemExpress
MedChemExpress IDO-IN-5 (NLG-1489) is an indoleamine 2,3-dioxygenase (IDO) inhibitor extracted from patent WO WO2012142237A1, compound 1489, has an IC50 of 1-10 μM.