CAS RN 1402709-93-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CCT241736, MedChemExpress
MedChemExpress CCT241736 is a potent and orally bioavailable dual FLT3 and Aurora kinase inhibitor, which inhibits Aurora kinases (Aurora-A Kd, 7.5 nM, IC50, 38 nM; Aurora-B Kd, 48 nM), FLT3 kinase (Kd, 6.2 nM), and FLT3 mutants including FLT3-ITD (Kd, 38 nM) and FLT3(D835Y) (Kd, 14 nM).