CAS RN 1402612-62-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ KT 182
Potent, selective and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor
2
KT182, MedChemExpress
MedChemExpress KT182 is a potent and selective inhibitor of α/β-hydrolase domain containing 6 (ABHD6), with an IC50 of 0.24 nM in Neuro2A cells.