CAS RN 1393465-84-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PLX7904, MedChemExpress
MedChemExpress PLX7904 is a potent and selective BRAF inhibitor, with IC50 of appr 5 nM against BRAFV600E in mutant RAS expressing cells.