CAS RN 138736-73-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HAMNO, MedChemExpress
MedChemExpress HAMNO is a novel protein interaction inhibitor of replication protein A (RPA).