CAS RN 1373765-19-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IPSU, MedChemExpress
MedChemExpress IPSU is a selective, orally available and brain penetrant OX2R antagonist with a pKi of 7.85.