CAS RN 136-16-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Oxythiamine, MedChemExpress
MedChemExpress Oxythiamine, an antimetabolite and a vitamin B1 antagonist, is a well-known thiamine antagonist and inhibitor of transketolase.