CAS RN 1357920-84-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Belizatinib, MedChemExpress
MedChemExpress Belizatinib is an oral, dual, potent inhibitor of ALK and TRKA, TRKB, and TRKC, with IC50 of 0.7 nM for wild-type recombinant ALK kinase.