CAS RN 1357350-60-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ALLO-2, MedChemExpress
MedChemExpress ALLO-2 is a potent drug-resistant Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with IC50 of 6 nM.