CAS RN 1346133-08-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LY2795050, MedChemExpress
MedChemExpress LY2795050 is a novel selective κ-opioid Receptor (KOR) antagonist (IC50=0.72 nM) and has the potential as a PET tracer to image KOR in vivo.