CAS RN 1338824-21-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ML224, MedChemExpress
MedChemExpress ML224(NCGC00242364; ANTAG3) is a selective TSHR inverse agonist; inhibits TSH-stimulated cAMP production with an IC50 = 2.3 μM.