CAS RN 1338466-77-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ EPZ 004777
Highly potent DOT1L inhibitor
2
EPZ004777, MedChemExpress
MedChemExpress EPZ004777 is a potent, selective DOT1L inhibitor with an IC50 of 0.4 nM.
3