CAS RN 133514-43-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Avexitide
MedChemExpress Avexitide (Exendin (9-39)) is a specific and competitive GLP-1 receptor antagonist.
2
MedChemExpress Avexitide
MedChemExpress Avexitide (Exendin (9-39)) is a specific and competitive GLP-1 receptor antagonist.